5-Methyl-2-(trifluormethyl)benzaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

H26411.06EA 524 EUR
H26411.06 H26411.03
5-Methyl-2-(trifluormethyl)benzaldehyd ≥97%
5-Methyl-2-(trifluormethyl)benzaldehyd
Formel: C₉H₇F₃O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06660294
CAS-Nummer: 886502-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR