2-Methoxy-N-methyanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL630926-1GEA 116 EUR
SIAL630926-1G
2-Methoxy-N-methyanilin, Sigma-Aldrich®
2-Methoxy-N-methyanilin
Formel: C₈H₁₁NO
MDL Number: MFCD03093822
CAS-Nummer: 10541-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR