2-Methylhippursäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL328006-5GEA
92.8
EUR
SIAL328006-5G
SIAL328006-1G
2-Methylhippursäure, Sigma-Aldrich®
2-Methylhippursäure
Formel:
C₁₀H₁₁NO₃ Molekulargewicht: 193,1992 g/mol |
MDL Number:
MFCD00050991 CAS-Nummer: 42013-20-7 EINECS: 255-622-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...