Vincristinsulfat
EDQMV0400000EA
235
EUR
EDQMV0400000
Vincristinsulfat
Vincristine sulphate
Formel:
C₄₆H₅₈N₄O₁₄S Molekulargewicht: 923,04 g/mol |
MDL Number:
MFCD00084729 CAS-Nummer: 2068-78-2 EINECS: 218-190-0 UN-Nummer: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...