(S)-(+)-2-Aminoheptan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL462632-1GEA 67.9 EUR
SIAL462632-1G
(S)-(+)-2-Aminoheptan, Sigma-Aldrich®
(S)-(+)-2-Aminoheptan
Formel: C₇H₁₇N
MDL Number: MFCD00066324
CAS-Nummer: 44745-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR