1,1,1,3,3,3-Hexafluorisopropylacrylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL367656-5GEA 116 EUR
SIAL367656-5G
1,1,1,3,3,3-Hexafluorisopropylacrylat, Sigma-Aldrich®
1,1,1,3,3,3-Hexafluorisopropylacrylat
Formel: C₆H₄F₆O₂
Molekulargewicht: 222,09 g/mol
MDL Number: MFCD00040104
CAS-Nummer: 2160-89-6
EINECS: 218-479-1
UN-Nummer: 3272
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR