1H,1H,3H-Hexafluorbutylacrylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL474452-25MLEA 340 EUR
SIAL474452-25ML
1H,1H,3H-Hexafluorbutylacrylat, Sigma-Aldrich®
1H,1H,3H-Hexafluorbutylacrylat
Formel: C₇H₆F₆O₂
MDL Number: MFCD00044091
CAS-Nummer: 54052-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR