Tetradecanophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL298611-5GEA 59.7 EUR
SIAL298611-5G
Tetradecanophenon, Sigma-Aldrich®
Tetradecanophenon
Formel: C₂₀H₃₂O
MDL Number: MFCD00008985
CAS-Nummer: 4497-05-6
EINECS: 224-790-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR