4'-Butoxyacetophenon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL540641-5GEA
87.5
EUR
SIAL540641-5G
4'-Butoxyacetophenon, Sigma-Aldrich®
4'-Butoxyacetophenon
Formel:
C₁₂H₁₆O₂ Molekulargewicht: 192,26 g/mol |
MDL Number:
MFCD00027200 CAS-Nummer: 5736-89-0 EINECS: 227-248-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...