Strontiumisopropoxid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL440698-25GEA
496
EUR
SIAL440698-25G
SIAL440698-5G
Strontiumisopropoxid, Sigma-Aldrich®
Strontiumisopropoxid
Formel:
C₆H₁₄O₂Sr Molekulargewicht: 205,79 g/mol |
MDL Number:
MFCD00050487 CAS-Nummer: 88863-33-6 UN-Nummer: 3205 ADR: 4.2,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...