Disulfoton

Lieferant: EHRENSTORFER GMBH

EHERCA12980000EA 89.3 EUR
EHERCA12980000
Disulfoton
Disulfoton
Formel: C₈H₁₉O₂PS₃
CAS-Nummer: 298-04-4
EINECS: 206-054-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR