(S)-(-)-Pulegon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL328847-1MLEA 319 EUR
SIAL328847-1ML
(S)-(-)-Pulegon, Sigma-Aldrich®
(S)-(-)-Pulegon
Formel: C₁₀H₁₆O
Molekulargewicht: 152,23 g/mol
MDL Number: MFCD00142347
CAS-Nummer: 3391-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR