4-(2-Pyridyl)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck

SIAL489387-5GEA 61.2 EUR
SIAL489387-5G
4-(2-Pyridyl)benzaldehyd, Sigma-Aldrich®
4-(2-Pyridyl)benzaldehyd
Formel: C₁₂H₉NO
Molekulargewicht: 183,21 g/mol
MDL Number: MFCD01863537
CAS-Nummer: 127406-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR