3-Amino-2-chlor-4-methylpyridin

Lieferant: LGC Standards PROMOCHEM

PROOMM1146.04EA 0 EUR
PROOMM1146.04
3-Amino-2-chlor-4-methylpyridin
3-Amino-2-chloro-4-methylpyridine
Formel: C₆H₇ClN₂
Molekulargewicht: 142,59 g/mol
MDL Number: MFCD00673152
CAS-Nummer: 133627-45-9
UN-Nummer: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR