Silver perrhenate, Sigma-Aldrich®

Lieferant: Merck

SIAL574198-1GEA 183 EUR
SIAL574198-1G
Silver perrhenate, Sigma-Aldrich®
Silver perrhenate
Formel: AgO₄Re
Molekulargewicht: 358,07 g/mol
MDL Number: MFCD00014144
CAS-Nummer: 20654-56-2
EINECS: 232-042-2
UN-Nummer: 1479
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR