Dishes, cylindrical
Lieferant: Avignon
AVIG10509EA 142 EUR
AVIG10509 AVIG10505 AVIG10507 AVIG10503 AVIG10510
Dishes, cylindrical
Made of porcelain.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean