Dodecacarbonyltetrairidium, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Iridium carbonyl

M-Clarity™ Qualitätsstufe = MQ100

299391-500MG
SIAL299391-500MGEA 207 EUR
SIAL299391-500MG
Dodecacarbonyltetrairidium, Sigma-Aldrich®
Dodecacarbonyltetrairidium
Formel: C₁₂Ir₄O₁₂
Molekulargewicht: 1104,99 g/mol
MDL Number: MFCD00011064
CAS-Nummer: 11065-24-0
UN-Nummer: 3281
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR