(4-Phthalimidobutyl)triphenylphosphoniumbromid ≥97%
|
Achtung
|
B22421.06EA
42.9
EUR
B22421.06
(4-Phthalimidobutyl)triphenylphosphoniumbromid ≥97%
(4-Phthalimidobutyl)triphenylphosphoniumbromid
Formel:
C₃₀H₂₇BrNO₂P Molekulargewicht: 544,43 g/mol Schmelzpunkt: 200…202 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02683092 CAS-Nummer: 65273-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...