D(+)-Altrose

Lieferant: Molekula

MOLE49386293-100MGEA 280 EUR
MOLE49386293-100MG
D(+)-Altrose
D(+)-Altrose
Formel: C₆H₁₂O₆
MDL Number: MFCD00075620
CAS-Nummer: 1990-29-0
EINECS: 217-870-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR