D(+)-Talose, Sigma-Aldrich®

Lieferant: Merck

SIAL86265-250MGEA 416 EUR
SIAL86265-250MG
D(+)-Talose, Sigma-Aldrich®
D(+)-Talose
Formel: C₆H₁₂O₆
Molekulargewicht: 180,16 g/mol
MDL Number: MFCD00135834
CAS-Nummer: 2595-98-4
EINECS: 219-996-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR