L(-)-Allose

Lieferant: Molekula

MOLEM75562029EA 0 EUR
MOLEM75562029
L(-)-Allose
L(-)-Allose
Formel: C₆H₁₂O₆
MDL Number: MFCD00085373
CAS-Nummer: 7635-11-2
EINECS: 231-565-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR