Benzyltriphenylphosphoniumtetrahydroborat

Lieferant: Molekula

MOLE76621546-5GEA 0 EUR
MOLE76621546-5G
Benzyltriphenylphosphoniumtetrahydroborat
Benzyltriphenylphosphoniumtetrahydroborat
Formel: C₂₅H₂₆BP
MDL Number: MFCD00800636
CAS-Nummer: 18117-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR