Benzyldimethyltetradecylammoniumchlorid Dihydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIAL292796-25GEA
159
EUR
SIAL292796-25G
Benzyldimethyltetradecylammoniumchlorid Dihydrat, Sigma-Aldrich®
Benzyldimethyltetradecylammoniumchlorid Dihydrat
Formel:
C₂₃H₄₂ClN·2H₂O Molekulargewicht: 404,0699 g/mol |
MDL Number:
MFCD00149985 CAS-Nummer: 147228-81-7 EINECS: 205-352-0 UN-Nummer: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...