Linalylacetat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ300
SIALW263613-1KG-KEA
494
EUR
SIALW263613-1KG-K
SIALW263613-100G-K
Linalylacetat, Sigma-Aldrich®
Linalylacetat
Formel:
C₁₂H₂₀O₂ Molekulargewicht: 196,2860 g/mol |
MDL Number:
MFCD00008907 CAS-Nummer: 115-95-7 EINECS: 204-116-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...