3-Chlor-5-fluorsalicylaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL593400-1GEA 205 EUR
SIAL593400-1G
3-Chlor-5-fluorsalicylaldehyd, Sigma-Aldrich®
3-Chlor-5-fluorsalicylaldehyd
Formel: C₇H₄ClFO₂
Molekulargewicht: 174,5569 g/mol
MDL Number: MFCD04039291
CAS-Nummer: 82128-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR