Methylarachidat
Lieferant: EHRENSTORFER GMBH
EHERC13112840EA
75.7
EUR
EHERC13112840
Methylarachidat
Methylarachidat
Formel:
C₂₁H₄₂O₂ Molekulargewicht: 326,56 g/mol |
MDL Number:
MFCD00009014 CAS-Nummer: 1120-28-1 EINECS: 214-304-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...