4-Hydroxy-2,6-dimethoxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL392936-1GEA 154 EUR
SIAL392936-1G
4-Hydroxy-2,6-dimethoxybenzaldehyd, Sigma-Aldrich®
4-Hydroxy-2,6-dimethoxybenzaldehyd
Formel: C₉H₁₀O₄
MDL Number: MFCD01407645
CAS-Nummer: 22080-96-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR