5-Chlor-2,3-dimethoxybenzaldehyd, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL571008-5GEA
462
EUR
SIAL571008-5G
5-Chlor-2,3-dimethoxybenzaldehyd, Sigma-Aldrich®
5-Chlor-2,3-dimethoxybenzaldehyd
Formel:
C₉H₉ClO₃ Molekulargewicht: 200,6200 g/mol |
MDL Number:
MFCD01743107 CAS-Nummer: 86232-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...