Methyl-4-phenylbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: p-Phenylbenzoic acid-OMe

SIALP5501-5GEA 96.5 EUR
SIALP5501-5G
Methyl-4-phenylbenzoat, Sigma-Aldrich®
Methyl-4-phenylbenzoat
Formel: C₁₄H₁₂O₂
Molekulargewicht: 212,25 g/mol
MDL Number: MFCD00017200
CAS-Nummer: 720-75-2
EINECS: 211-954-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR