(3-Fluorbenzyl)triphenylphosphoniumchlorid
Lieferant: Thermo Scientific
MAYBKM00117.1EA
0
EUR
MAYBKM00117.1
MAYBKM00117.250
(3-Fluorbenzyl)triphenylphosphoniumchlorid
(3-Fluorbenzyl)triphenylphosphoniumchlorid
Formel:
C₂₅H₂₁ClFP |
MDL Number:
MFCD00179768 CAS-Nummer: 55128-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...