Hafniumtetrakis(dimethylazanid), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
SIAL666610-25GEA
2660
EUR
SIAL666610-25G
Hafniumtetrakis(dimethylazanid), Sigma-Aldrich®
Tetrakis(dimethylamino)hafnium(IV)
Formel:
C₈H₂₄HfN₄ Molekulargewicht: 354,793 g/mol |
MDL Number:
MFCD01862473 CAS-Nummer: 19782-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...