Tube rotator, Rotamix
Lieferant: Bellco Glass
BELC7770-10230EA 3740 EUR
BELC7770-10230
Tube rotator, Rotamix
Schüttler Rotations-/Wippschüttler
Rotamix tube rotator with digital display provides 360° rotation for all types of mixing and extraction protocols.
  • Small compact design and variable speed control with an LCD digital display
  • Ideal for immunoprecipitation extractions and phase separations
  • Can be used in the cold room, at room temperature or in an incubator
  • Chemical resistant coated steel case, stainless steel rods (available separately)
Bestellinformation: Interchangeable rods need to be ordered separately.