Protection cover for poly IBC spill containment pallets, PIG®
Lieferant: New Pig
NEWPPAK753EA 0 EUR
NEWPPAK753
Protection cover for poly IBC spill containment pallets, PIG®
This cover for outdoor storage is designed for the protection of poly IBC spill containment pallets.
  • Prevents rainwater and snowmelt from overflowing the poly IBC spill containment pallet
  • Elasticised bottom creates a tight fit around IBC base to divert leaks into sump
  • Clear materials enable users to check the sumps without removing the cover
  • Made of 4-mil anti-static PE to help dissipate static charges