1,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
Lieferant: Thermo Scientific
MAYBMAY00074DAEA
0
EUR
MAYBMAY00074DA
1,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
1,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
Formel:
C₁₁H₁₉BN₂O₂ Molekulargewicht: 222,0918 g/mol |
MDL Number:
MFCD09864189 CAS-Nummer: 1036991-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...