(1R)-(-)-Campher-10-sulfonamid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL375632-1GEA
59.6
EUR
SIAL375632-1G
(1R)-(-)-Campher-10-sulfonamid, Sigma-Aldrich®
(1R)-(-)-Campher-10-sulfonamid
Formel:
C₁₀H₁₇NO₃S Molekulargewicht: 231,32 g/mol |
MDL Number:
MFCD00151500 CAS-Nummer: 72597-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...