Histamin Dihydrochlorid

Lieferant: Enzo Life Sciences

ALEXALX550132G005EA 68.6 EUR
ALEXALX550132G005
Histamin Dihydrochlorid
Histamin Dihydrochlorid
Formel: C₅H₁₁Cl₂N₃
Molekulargewicht: 184,07 g/mol
MDL Number: MFCD00012703
CAS-Nummer: 56-92-8
EINECS: 200-298-4
https://at.vwr.com/stibo/web/std.lang.all/08/10/45390810.jpg

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR