(3,4-Dichlorphenyl)methansulfonsäurechlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL678627-500MGEA 119 EUR
SIAL678627-500MG
(3,4-Dichlorphenyl)methansulfonsäurechlorid, Sigma-Aldrich®
(3,4-Dichlorophenyl)methanesulphonyl chloride
Formel: C₇H₅CI₃O₂S
MDL Number: MFCD01631929
CAS-Nummer: 85952-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR