1-Fluor-2,4,6-trimethylpyridiniumtrifluormethansulfonat, Sigma-Aldrich®


M-Clarity™ quality level = MQ100

SIAL378216-1GEA 110 EUR
SIAL378216-1G
1-Fluor-2,4,6-trimethylpyridiniumtrifluormethansulfonat, Sigma-Aldrich®
1-Fluoro-2,4,6-trimethylpyridiniumtrifluoromethanesulphonate
Formel: C₉H₁₁F₄NO₃S
MDL Number: MFCD00067525
CAS-Nummer: 107264-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR