3-Ethoxy-2,6-difluoranilin ≥97%, JRD
|
Gefahr
|
H32732.03EA
99.3
EUR
H32732.03
H32732.06
3-Ethoxy-2,6-difluoranilin ≥97%, JRD
3-Ethoxy-2,6-difluoranilin
Formel:
C₈H₉F₂NO Molekulargewicht: 173,16 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11519309 CAS-Nummer: 651734-64-4 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...