Kokosnussöl

Lieferant: Merck

46949.EA 84.1 EUR
46949.
Kokosnussöl
Coconut oil
MDL Number: MFCD00130817
CAS-Nummer: 8001-31-8
EINECS: 232-282-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR