(3-Fluorphenyl)methansulfonsäurechlorid

MAYBMO07830DAEA 0 EUR
MAYBMO07830DA
(3-Fluorphenyl)methansulfonsäurechlorid
(3-Fluorophenyl)methanesulphonyl chloride
Formel: C₇H₆ClFO₂S
MDL Number: MFCD04117456
CAS-Nummer: 24974-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR