Methyl-3-methyl-4-nitrobenzoat
Lieferant: Thermo Scientific
MAYBTL00715EEEA
0
EUR
MAYBTL00715EE
MAYBTL00715DA
Methyl-3-methyl-4-nitrobenzoat
Methyl-3-methyl-4-nitrobenzoat
Formel:
C₉H₉NO₄ Molekulargewicht: 195,18 g/mol |
MDL Number:
MFCD00085640 CAS-Nummer: 24078-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...