Methyl-2,5-dibrombenzoat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL532894-25GEA 156 EUR
SIAL532894-25G
Methyl-2,5-dibrombenzoat, Sigma-Aldrich®
Methyl-2,5-dibrombenzoat
Formel: C₈H₆Br₂O₂
Molekulargewicht: 293,94 g/mol
MDL Number: MFCD00144757
CAS-Nummer: 57381-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR