Spatulas, Azlon®
Lieferant: SciLabware
Azlon®
231-0096EA 0 EUR
231-0096
Spatulas, Azlon®
Spatel
Moulded strong tapered spatulas with chamfered ends. Made of translucent PP.
  • Virtually unbreakable in normal use and retain shape after flexing
  • Good chemical and heat resistance
  • Autoclavable at 121 °C