prepGEM® DNA extraction kits have been developed to provide a rapid, reliable, and economical solution to DNA extraction. The kits use a single tube protocol with temperature control to prepare DNA for analysis, and are optimised for downstream PCR-based applications, including qPCR, STR-PCR, DNA sequencing, AFLP analysis, and many SNiP detection applications.
prepGEM® Storage Card Saliva kit is specifically developed for extracting DNA from human saliva stored on FTA cards. The kit is adapted from Saliva kit, and optimised to overcome the PCR inhibitors present in some brands of storage card. The extracted DNA is suitable for PCR applications including qPCR, STR-PCR detection, and other genotyping applications.