Nickel(II)tetrafluoroborat 45% in wässriger Lösung
SIAL01587-40KG-HEA
539
EUR
SIAL01587-40KG-H
Nickel(II)tetrafluoroborat 45% in wässriger Lösung
Nickel(II) tetrafluoroborate
Formel:
B₂F₈Ni |
EINECS:
238-753-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...