2-(4-Bromphenyl)imidazo[1,2-a]pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL658979-1GEA 35.7 EUR
SIAL658979-1G
2-(4-Bromphenyl)imidazo[1,2-a]pyridin, Sigma-Aldrich®
2-(4-Bromphenyl)imidazo[1,2-a]pyridin
Formel: C₁₃H₉BrN₂
Molekulargewicht: 273,13 g/mol
MDL Number: MFCD00218250
CAS-Nummer: 34658-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR