3-Brom-1-methyl-1H-indazol, Sigma-Aldrich®

Lieferant: Merck

SIAL717215-1GEA 103 EUR
SIAL717215-1G
3-Brom-1-methyl-1H-indazol, Sigma-Aldrich®
3-Bromo-1-methyl-1H-indazole
Formel: C₈H₇BrN₂
Molekulargewicht: 211,0586 g/mol
MDL Number: MFCD12923368
CAS-Nummer: 326474-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR