4-(Chlormethyl)-3-(2,6-dichlorphenyl)-5-methylisoxazol
Lieferant: Thermo Scientific
MAYBSPB07543.1EA
0
EUR
MAYBSPB07543.1
4-(Chlormethyl)-3-(2,6-dichlorphenyl)-5-methylisoxazol
4-(Chloromethyl)-3-(2,6-dichlorophenyl)-5-methylisoxazole
Formel:
C₁₁H₈Cl₃NO |
MDL Number:
MFCD00833265 CAS-Nummer: 303225-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...