Okadasäure Ammoniumsalz, Millipore®
Lieferant: Merck Millipore Calbiochem
459616-25EA
99.3
EUR
459616-25
Okadasäure Ammoniumsalz, Millipore®
Okadaic acid ammonium salt
Formel:
C₄₄H₆₇O₁₃·NH₄ Molekulargewicht: 822,03 g/mol |
MDL Number:
MFCD00153856 CAS-Nummer: 155716-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...